Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Phosphate pH 7.5 DILUT-IT™ Dissolution Media Concentrate, J.T.Baker™

Click to view available options
Quantity:
230.8mL
961.5mL
Packaging:
Narrow Mouth Polyethylene Bottle
Poly Bottle

Chemical Identifiers

Potassium Phosphate pH 7.5 DILUT-IT™ Dissolution Media Concentrate
Packaging Narrow Mouth Polyethylene Bottle
Specific Gravity 1L = 1.00kg
Quantity 961.5mL
Chemical Name or Material Potassium Phosphate pH 7.5 DILUT-IT™ Dissolution Media Concentrate