Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Phosphate, Tribasic, n-Hydrate, Powder, BAKER ANALYZED™ Reagent, J.T.Baker™

Click to view available options
Quantity:
2.5kg
500g
Packaging:
Poly Bottle

Chemical Identifiers

7778-53-2
K3O4P
212.265
LWIHDJKSTIGBAC-UHFFFAOYSA-K
62657

High quality chemicals for laboratory and specialized industrial use

CAS 7778-53-2
Molecular Formula K3O4P
InChI Key LWIHDJKSTIGBAC-UHFFFAOYSA-K
SMILES [O-]P(=O)([O-])[O-].[K+].[K+].[K+]
IUPAC Name tripotassium;phosphate
Molecular Weight (g/mol) 212.265
PubChem CID 62657
Physical Form Powder
Packaging Poly Bottle
Quantity 500g
Chemical Name or Material Potassium Phosphate, Tribasic, n-Hydrate
Grade BAKER ANALYZED™ Reagent
Show More Show Less