Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Phosphate USP GenAR, Macron™

Click to view available options
Quantity:
500g
4 x 2.5kg
Packaging:
POLYSTORMOR
Poly
7758-11-4
HK2O4P
174.17
ZPWVASYFFYYZEW-UHFFFAOYSA-L
dipotassium hydrogen phosphate, dipotassium phosphate, dipotassium hydrogenphosphate, dibasic potassium phosphate, potassium hydrogen phosphate, potassium phosphate, dibasic, potassium dibasic phosphate, potassium phosphate dibasic, phosphoric acid, dipotassium salt, dipotassium monophosphate
24450
CHEBI:32031
dipotassium;hydrogen phosphate
OP(=O)([O-])[O-].[K+].[K+]
7758-11-4
HK2O4P
174.17
ZPWVASYFFYYZEW-UHFFFAOYSA-L
dipotassium hydrogen phosphate, dipotassium phosphate, dipotassium hydrogenphosphate, dibasic potassium phosphate, potassium hydrogen phosphate, potassium phosphate, dibasic, potassium dibasic phosphate, potassium phosphate dibasic, phosphoric acid, dipotassium salt, dipotassium monophosphate
24450
CHEBI:32031
dipotassium;hydrogen phosphate
OP(=O)([O-])[O-].[K+].[K+]
Formula Weight 174.18
Grade USP
Packaging Poly
Quantity 4 x 2.5kg
Chemical Name or Material Potassium Phosphate, Anhydrous, Dibasic