Potassium plasma standard solution, Alfa Aesar™, Specpure™

Manufacturer: Alfa Aesar 13866AE

Quantity 100mL

 View more versions of this product

Catalog No. AA13866AE

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7697-37-2,7757-79-1,7732-18-5
    Physical Form Liquid
    Molecular Formula KNO3 in 5% HNO3
    MDL Number MFCD00133776
    Chemical Name or Material Potassium

    Specifications

    Color Clear colorless
    Physical Form Liquid
    Quantity 100mL
    DOT Information Hazard Class: 8, Packing Group: III
    EINECS Number 231-714-2
    Appearance Clear liquid
    Molecular Formula KNO3 in 5% HNO3
    CAS 7697-37-2,7757-79-1,7732-18-5
    Concentration or Composition (by Analyte or Components) Nitric acid: 5.0%; Potassium nitrate: 0.26%; Water: 94.74%
    MDL Number MFCD00133776
    Health Hazard 1 H314: Causes severe skin burns and eye damage.
    Vapor Pressure 23 hPa (17mm Hg) at 20°C
    Chemical Name or Material Potassium

    H314: Causes severe skin burns and eye damage.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit