Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium polysulfide, pure, ACROS Organics™

Click to view available options
Quantity:
250g
1kg
2.5kg
Packaging:
Plastic bottle
37199-66-9
H2KS+
73.174
MFCD00148074
ZOCLAPYLSUCOGI-UHFFFAOYSA-N
potassium sulfane,potassium polysulfide,caswell no. 701,potassium sulfide k2 sx,epa pesticide chemical code 076703
5362529
potassium;sulfane
S.[K+]
37199-66-9
H2KS+
73.174
MFCD00148074
ZOCLAPYLSUCOGI-UHFFFAOYSA-N
potassium sulfane,potassium polysulfide,caswell no. 701,potassium sulfide k2 sx,epa pesticide chemical code 076703
5362529
potassium;sulfane
S.[K+]
Linear Formula (K2S)n
Solubility Information Solubility in water: 500g/l (20°C)
Grade Pure
Packaging Plastic bottle
Solubility Solubility in water: 500g/l (20°C).
Melting Point 200.0°C to 250.0°C
Quantity 2.5kg
Chemical Name or Material Potassium polysulfide

GHS H Statement:
Causes severe skin burns and eye damage.
Very toxic to aquatic life.
Contact with acids liberates toxic gas.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 253-390-1