Alfa Aesar™ Potassium pyrophosphate, 95%

Manufacturer: Alfa Aesar™ 13436A7

Quantity 5kg

 View more versions of this product

Catalog No. AA13436A7

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7320-34-5
    Formula Weight 330.34
    MDL Number MFCD00011393
    Merck Index 14,7663
    Chemical Name or Material Potassium pyrophosphate

    Specifications

    Formula Weight 330.34
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Sensitivity Hygroscopic
    MDL Number MFCD00011393
    Melting Point 1090°C
    Health Hazard 1 Warning
    TSCA TSCA
    Physical Form Powder
    Merck Index 14,7663
    Solubility Information Soluble in water
    Density 2.33g/mL
    CAS 7320-34-5
    Color White
    Quantity 5kg
    Chemical Name or Material Potassium pyrophosphate
    Assay 95%

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit