Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Pyrophosphate, Powder, FCC, 95%, Spectrum™

Click to view available options
Quantity:
500 g
2.5 kg
12 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
7320-34-5
K4O7P2
330.33
RYCLIXPGLDDLTM-UHFFFAOYSA-J
tetrapotassium (phosphonatooxy)phosphonate
[K+].[K+].[K+].[K+].[O-]P([O-])(=O)OP([O-])([O-])=O
7320-34-5
K4O7P2
330.33
RYCLIXPGLDDLTM-UHFFFAOYSA-J
tetrapotassium (phosphonatooxy)phosphonate
[K+].[K+].[K+].[K+].[O-]P([O-])(=O)OP([O-])([O-])=O

P1462, 7320-34-5, K4P2O7

Spectrum™ Potassium Pyrophosphate, Powder, FCC is used as a a sequestering, buffering and emulsifying agent. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage, nutritional supplement and personal care applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

CAS Max % 1
Arsenic (As) 3 mg/kg
Percent Purity 95%
Grade FCC
Fluoride 10 mg/kg
Insoluble Matter 0.001
Lead (Pb) 2 mg/kg
Loss on Ignition 0.005
Packaging Poly Bottle
Quantity 500 g