Potassium selenate, 99.5% (metals basis), Alfa Aesar™

Manufacturer: Alfa Aesar 6811109

Quantity 10g

 View more versions of this product

Catalog No. AA6811109

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7790-59-2
    Physical Form Powder
    Assay Percent Range 99.5% (metals basis)
    Molecular Formula K2SeO4
    Formula Weight 221.16
    MDL Number MFCD00054125
    Chemical Name or Material Potassium selenate

    Specifications

    Physical Form Powder
    Formula Weight 221.16
    Quantity 10g
    Color White
    Density 3.07g/cm3 at 20°C
    EINECS Number 232-214-7
    Odor Odorless
    Molecular Formula K2SeO4
    CAS 7790-59-2
    Assay Percent Range 99.5% (metals basis)
    MDL Number MFCD00054125
    Health Hazard 1 H301-H331-H373-H400-H410: Toxic if swallowed. Toxic if inhaled. May cause damage to organs through prolonged or repeated exposure. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects.
    Chemical Name or Material Potassium selenate

    H301-H331-H373-H400-H410: Toxic if swallowed. Toxic if inhaled. May cause damage to organs through prolonged or repeated exposure. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit