Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium selenocyanate, 98.5%, Thermo Scientific Chemicals

Catalog Number AA6810718
Click to view available options
Quantity:
10 g
50 g
3425-46-5
CKNSe
144.087
MFCD00011366
KYEKHFSRAXRJBR-UHFFFAOYSA-M
potassium selenocyanate, selenocyanic acid, potassium salt, potassium selenoisocyanate, ccris 7063, potassium ion selenocyanate, potassium selenocyanate 10g
76960
potassium;selenocyanate
C(#N)[Se-].[K+]
3425-46-5
CKNSe
144.087
MFCD00011366
KYEKHFSRAXRJBR-UHFFFAOYSA-M
potassium selenocyanate, selenocyanic acid, potassium salt, potassium selenoisocyanate, ccris 7063, potassium ion selenocyanate, potassium selenocyanate 10g
76960
potassium;selenocyanate
C(#N)[Se-].[K+]

Potassium selenocyanate is used to prepare triphenylphosphine selenide by reacting with triphenylphosphine. It finds application in solid state NMR of heavy-metal nuclei which results in the preparation of several mercury thiocyanate and selenocyanate compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 162°C to 164°C
Color White
Physical Form Crystalline
Quantity 50 g
UN Number UN3283
Solubility Information Soluble in water,ethanol,dimethyl formamide,hexamethylphosphoramide,acetonitrile and methanol. Slightly soluble in terahydrofuran.
Formula Weight 144.08
Percent Purity 98.5%
Odor Odorless
Sensitivity Moisture sensitive
Density 2.347 g/mL
Chemical Name or Material Potassium selenocyanate
Show More Show Less
Hazard Category H301+H331-H373
Hazard Statement GHS H Statement
H301-H331-H373
Toxic if swallowed.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Precautionary Statement P260-P264b-P270-P271-P301+P310-P304+P340-P311-P314-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: SELENIUM COMPOUND, SOLID, N.O.S.
EINECSNumber 222-320-1
RTECSNumber VT4000000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit