Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium selenocyanate, 99%, ACROS Organics™

Click to view available options
Quantity:
10g
50g
250g
Packaging:
Glass bottle
3425-46-5
CKNSe
144.08
MFCD00011366
KYEKHFSRAXRJBR-UHFFFAOYSA-M
potassium selenocyanate,selenocyanic acid, potassium salt,potassium selenoisocyanate,ccris 7063,potassium ion selenocyanate,potassium selenocyanate 10g
76960
potassium;selenocyanate
C(#N)[Se-].[K+]
3425-46-5
CKNSe
144.08
MFCD00011366
KYEKHFSRAXRJBR-UHFFFAOYSA-M
potassium selenocyanate,selenocyanic acid, potassium salt,potassium selenoisocyanate,ccris 7063,potassium ion selenocyanate,potassium selenocyanate 10g
76960
potassium;selenocyanate
C(#N)[Se-].[K+]
Assay Percent Range 98.5 % min. (Argentometry)
Linear Formula KSeCN
Fieser 06,487; 08,417; 13,186; 08,417; 15,274
Solubility Information Solubility in water: soluble
Formula Weight 144.08
Physical Form Crystalline Solid, Powder, Crystals and/or Chunks
Percent Purity 99%
Infrared Spectrum Authentic
Packaging Glass bottle
Solubility Solubility in water: soluble.
Color Orange to Red
Melting Point 152.0°C to 158.0°C
Quantity 250g
Chemical Name or Material Potassium selenocyanate
Show More Show Less

GHS H Statement
Toxic if swallowed.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure if inhaled.
Very toxic to aquatic life with long lasting effects.

GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician.
Store in a well-

GHS Signal Word: Danger

EINECSNumber : 222-320-1