Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Potassium sodium L-tartrate tetrahydrate, 99%

Manufacturer:  Alfa Aesar™ A101630I

Quantity 5000g

 View more versions of this product

Catalog No. AAA101630I


  Check Availability
Add to cart

Specifications

Description

Applications
Baking powder, medicine (cathartic), component of Fehling ft.s solution, silvering mirrors.Potassium sodium L-tartrate tetrahydrate is used in the process of silvering mirrors. It is active ingredient in the Fehiling’s solution and Nylander’s test involved in the determination of reducing sugars. It is utilized to break up emulsion in organic synthesis as well as a common precipitant in protein crystallography. As a Biuret reagent, it is used to measure the protein concentration. Furthermore, it is used as laxative and is also used in food industry.

Solubility
Soluble in water. Insoluble in alcohol

Notes
Loses water of crystallization at 140°. Unstable above 225°. Incompatible with strong oxidizing agents.
Description & Specifications

Chemical Identifiers

6381-59-5
282.23 (210.29 Anhydrous)
6113568
Rochelle Salt; Sodium potassium L-tartrate tetrahydrate
C4H4KNaO6·4H2O
MFCD00150989
14,7670
Potassium sodium L-tartrate tetrahydrate

Specifications

6381-59-5
C4H4KNaO6·4H2O
MFCD00150989
14,7670
Soluble in water; Insoluble in alcohol
70° to 80°C
5000g
Potassium sodium L-tartrate tetrahydrate
99%
282.23 (210.29 Anhydrous)
6113568
Rochelle Salt; Sodium potassium L-tartrate tetrahydrate
Odorless
6.0 to 8.5
+21° (c=1 in water)
99%

Safety and Handling

SDS & Safety Documents

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit