Learn More
Potassium sodium L-tartrate tetrahydrate, ACS, 99.0-102.0%, Thermo Scientific Chemicals
Description
Potassium sodium L-tartrate tetrahydrate is used in the process of silvering mirrors. It is an active ingredient in Fehiling’s solution and Nylander’s test involved in the determination of reducing sugars. It is utilized to break up emulsion in organic synthesis as well as a common precipitant in protein crystallography. As a Biuret reagent, it is used to measure the protein concentration. Furthermore, it is used as a laxative and is also used in food industry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 70°C to 80°C |
Color | Colorless |
Physical Form | Crystalline |
Quantity | 2.5 kg |
Assay Percent Range | 99.0-102.0% |
Beilstein | 6113568 |
Solubility Information | Soluble in water. Insoluble in alcohol. |
Formula Weight | 282.23 (210.29 Anhydrous) |
Grade | ACS |
Odor | Odorless |
Show More |
Safety and Handling
EINECSNumber | 206-156-8 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.