Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Sodium Tartrate, 4-Hydrate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
1kg
2.5kg
500g
Packaging:
Poly Bottle

Chemical Identifiers

6381-59-5
C4H12KNaO10
282.218
VZOPRCCTKLAGPN-UHFFFAOYSA-L
2724148

High quality chemicals for laboratory and specialized industrial use

CAS 6381-59-5
Molecular Formula C4H12KNaO10
InChI Key VZOPRCCTKLAGPN-UHFFFAOYSA-L
SMILES C(C(C(=O)[O-])O)(C(=O)[O-])O.O.O.O.O.[Na+].[K+]
IUPAC Name potassium;sodium;2,3-dihydroxybutanedioate;tetrahydrate
Molecular Weight (g/mol) 282.218
PubChem CID 2724148
Formula Weight 282.22
Physical Form Crystals
Packaging Poly Bottle
Specific Gravity 1L = 1.79kg
Quantity 500g
Chemical Name or Material Potassium Sodium Tartrate, 4-Hydrate
Grade Baker Analyzed™ ACS Reagent
Show More Show Less