Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Potassium sodium tartrate tetrahydrate, 99+%, for analysis, ACROS Organics™
Click to view available options
Quantity:
500g
1kg
5kg
25kg
Packaging:
Fibre drum
Plastic bottle
Specifications
Chloride | 50ppm max |
Sulfate | 50ppm max |
Specific Rotation | + 22.10 |
Packaging | Fibre drum |
Infrared Spectrum | Authentic |
Melting Point | 70.0°C to 80.0°C |
Quantity | 25kg |
Color | White |
pH | 5.5 to 8.5 (5% soln. at 20°C) |
Assay Percent Range | 99+% |
Show More |