Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Sodium Tartrate Tetrahydrate, Purified (Rochelle Salt), Reagents

Catalog No. p-200021236
Click to view available options
Quantity:
500 g
3 kg

Potassium Sodium Tartrate Tetrahydrate, Purified (Rochelle Salt), Chemically Pure, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Potassium Sodium Tartrate Tetrahydrate, Purified (Rochelle Salt)
Boiling Point 220°C
CAS 6381-59-5
Color Colorless/White
Physical Form Solid
Quantity 3 kg
Molecular Formula KNaC4H4O6, 4H2O
Synonym Rochelle Salt, Sodium potassium tartrate tetrahydrate
Molecular Weight (g/mol) 282.2201
Grade Pure
Packaging Poly Bottle
Specific Gravity 1.79
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.