Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium, solid, 99.95% (metals basis), Alfa Aesar™

Click to view available options
Quantity:
1g
5g
7440-09-7
K
39.10
MFCD00133776
NPYPAHLBTDXSSS-UHFFFAOYSA-N
kalium,potassium, metal,potassium, liquid alloy,unii-rwp5ga015d,rwp5ga015d,hsdb 698,monopotassium,potasio,mono-potassium,atom
5462222
CHEBI:26216
potassium
[K]
7440-09-7
K
39.10
MFCD00133776
NPYPAHLBTDXSSS-UHFFFAOYSA-N
kalium,potassium, metal,potassium, liquid alloy,unii-rwp5ga015d,rwp5ga015d,hsdb 698,monopotassium,potasio,mono-potassium,atom
5462222
CHEBI:26216
potassium
[K]
Quantity 1g
Appearance Silver colored solid
Assay Percent Range 99.95%
Odor Odorless
Physical Form Sealed in standard ampoules under vacuum
Color Silver
Chemical Name or Material Potassium

P223-P231+P232-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P335+P334-P363-P370+P378q-P501c

H260-H314-H335

DOTInformation : DOT Class: 4.3, Packaging Group: I

EINECSNumber : 231-119-8

RTECSNumber : TS6460000

TSCA : Yes

Recommended Storage : Ambient temperatures