Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Sorbate, Powder, NF, 98-101%, Spectrum™

Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
45 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
590-00-1
C6H7KO2
150.22
CHHHXKFHOYLYRE-STWYSWDKSA-M
potassium (2E,4E)-hexa-2,4-dienoate
[K+].C\C=C\C=C\C([O-])=O
590-00-1
C6H7KO2
150.22
CHHHXKFHOYLYRE-STWYSWDKSA-M
potassium (2E,4E)-hexa-2,4-dienoate
[K+].C\C=C\C=C\C([O-])=O

PO300, 590-00-1, C6H7KO2

Spectrum™ Potassium Sorbate, Powder, NF is used as an antimicrobial preservative. All Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Acidity 1.1 mL NaOH
CAS Max % 1
Alkalinity 0.80 mL HCl
Percent Purity 98 to 101%
Loss on Drying 0.01
Packaging Poly Pail
Quantity 2.5 kg