Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell Fluka™ Potassium Sulfate, ≥99.0%, ACS Reagent, Honeywell™ Fluka™

Click to view available options
Quantity:
500g
2.5kg
Packaging:
Poly Bottle
7778-80-5
K2O4S
174.253
MFCD00011388
OTYBMLCTZGSZBG-UHFFFAOYSA-L
potassium sulfate,dipotassium sulfate,potassium sulphate,sulfuric acid dipotassium salt,sal polychrestum,sulfuric acid, potassium salt,arcanum duplicatum,kalium sulphuricum,caswell no. 702,sulfuric acid, dipotassium salt
24507
CHEBI:32036
dipotassium;sulfate
[O-]S(=O)(=O)[O-].[K+].[K+]
7778-80-5
K2O4S
174.253
MFCD00011388
OTYBMLCTZGSZBG-UHFFFAOYSA-L
potassium sulfate,dipotassium sulfate,potassium sulphate,sulfuric acid dipotassium salt,sal polychrestum,sulfuric acid, potassium salt,arcanum duplicatum,kalium sulphuricum,caswell no. 702,sulfuric acid, dipotassium salt
24507
CHEBI:32036
dipotassium;sulfate
[O-]S(=O)(=O)[O-].[K+].[K+]

ACS Reagent,≥99.0%, powder

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Linear Formula K2SO4
UN Number NONH for all modes of transport
Appearance Powder
Formula Weight 174.26g/mol
Physical Form Powder
Percent Purity ≥99.0%
Grade ACS Reagent
Packaging Poly Bottle
pH Range 6.0 to 7.5 at 20°C
Solubility in Water 111g/L at 20°C
Boiling Point 1698°C
Melting Point 1067°C
Quantity 2.5kg
Chemical Name or Material Potassium Sulfate
Show More Show Less

EINECSNumber : 231-915-5

RTECSNumber : TT5900000