Potassium sulfate, ACS, 99.0% min, Alfa Aesar™

Manufacturer: Alfa Aesar 1431136

Quantity 500g

 View more versions of this product

Catalog No. AA1431136

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7778-80-5
    Physical Form Crystalline
    Assay Percent Range 99.0% min
    Molecular Formula K2SO4
    Formula Weight 174.27
    MDL Number MFCD00011388
    Chemical Name or Material Potassium sulfate, For ACS analysis

    Specifications

    Formula Weight 174.27
    Quantity 500g
    EINECS Number 231-915-5
    Physical Form Crystalline
    Density 2.662g/mL
    MDL Number MFCD00011388
    CAS 7778-80-5
    Assay Percent Range 99.0% min
    Molecular Formula K2SO4
    Chemical Name or Material Potassium sulfate, For ACS analysis

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit