Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Sulfate, BiotechGrade, 99%, Spectrum™

Click to view available options
Quantity:
500 g
6 x 500 g
Packaging:
Poly Bottle
7778-80-5
K2O4S
174.25
OTYBMLCTZGSZBG-UHFFFAOYSA-L
dipotassium sulfate
[K+].[K+].[O-]S([O-])(=O)=O
7778-80-5
K2O4S
174.25
OTYBMLCTZGSZBG-UHFFFAOYSA-L
dipotassium sulfate
[K+].[K+].[O-]S([O-])(=O)=O

P1409, 7778-80-5, K2SO4

Spectrum™ Potassium Sulfate, BiotechGrade , also known as sulphate of potash, is most often used in fertilizers which provides both sulfur and potassium. Spectrum offers highly pure reagents suitable for biochemical research and analysis. The critical parameters involved are absence of inhibitors such as traces of heavy metals as well as biochemical function tests for enzymes, coenzymes and enzyme substrates.

CAS Max % 1
Aluminum (Al) 5e-006
Arsenic (As) 1e-006
Percent Purity 99%
Grade Biotech
Calcium (Ca) 5e-005
Copper (Cu) 5e-006
Insoluble Matter 0.0001
Iron (Fe) 5e-006
Lead (Pb) 5e-006
Magnesium (Mg) 2e-005
Nitrogen Compounds 5 ppm
Packaging Poly Bottle
Sodium (Na) 5e-005
Water 0.01
Zinc (Zn) 5e-006
pH 5.5 to 8.5
Quantity 500 g
Show More Show Less