Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Sulfate, Low Nitrogen, Reagents

Catalog No. 01337087
Change view
Click to view available options
Quantity:
500 g
3 kg
12 kg
Catalog No. Quantity
01-337-087 3 kg
01-337-088 12 kg
01-337-086 500 g
3 options

Catalog No. 01-337-087

Supplier: Reagents Holdings Llc C22910003A5

Only null left
Add to Cart
Add to Cart

Potassium Sulfate, Low Nitrogen, Chemically Pure, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7778-80-5
Molecular Formula K2O4S
Molecular Weight (g/mol) 174.25
MDL Number MFCD00011388
InChI Key OTYBMLCTZGSZBG-UHFFFAOYSA-L
Synonym Dipotassium Sulfate
IUPAC Name dipotassium sulfate
SMILES [K+].[K+].[O-]S([O-])(=O)=O

Specifications

Boiling Point 1689°C
Melting Point 1069°C
Color Colorless/White
Physical Form Solid
Quantity 3 kg
Grade Pure
Packaging Poly Bottle
Specific Gravity 2.66
Chemical Name or Material Potassium Sulfate, Low Nitrogen
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.