Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Sulfate, Powder, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
2.5kg
500g
Packaging:
Poly Bottle

Chemical Identifiers

7778-80-5
K2O4S
174.253
OTYBMLCTZGSZBG-UHFFFAOYSA-L
24507
CHEBI:32036

High quality chemicals for laboratory and specialized industrial use

CAS 7778-80-5
Molecular Formula K2O4S
InChI Key OTYBMLCTZGSZBG-UHFFFAOYSA-L
SMILES [O-]S(=O)(=O)[O-].[K+].[K+]
IUPAC Name dipotassium;sulfate
Molecular Weight (g/mol) 174.253
PubChem CID 24507
ChEBI CHEBI:32036
Formula Weight 174.27
Physical Form Powder
Packaging Poly Bottle
Specific Gravity 1L = 2.66kg
Quantity 500g
Chemical Name or Material Potassium Sulfate
Grade Baker Analyzed™ ACS Reagent
Show More Show Less