Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Sulfate, Powder, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
500g
2.5kg
Packaging:
Poly Bottle
7778-80-5
K2O4S
174.253
OTYBMLCTZGSZBG-UHFFFAOYSA-L
potassium sulfate,dipotassium sulfate,potassium sulphate,sulfuric acid dipotassium salt,sal polychrestum,sulfuric acid, potassium salt,arcanum duplicatum,kalium sulphuricum,caswell no. 702,sulfuric acid, dipotassium salt
24507
CHEBI:32036
dipotassium;sulfate
[O-]S(=O)(=O)[O-].[K+].[K+]
7778-80-5
K2O4S
174.253
OTYBMLCTZGSZBG-UHFFFAOYSA-L
potassium sulfate,dipotassium sulfate,potassium sulphate,sulfuric acid dipotassium salt,sal polychrestum,sulfuric acid, potassium salt,arcanum duplicatum,kalium sulphuricum,caswell no. 702,sulfuric acid, dipotassium salt
24507
CHEBI:32036
dipotassium;sulfate
[O-]S(=O)(=O)[O-].[K+].[K+]

High quality chemicals for laboratory and specialized industrial use

Formula Weight 174.27
Physical Form Powder
Grade BAKER ANALYZED™ ACS Reagent
Packaging Poly Bottle
Specific Gravity 1L = 2.66kg
Quantity 2.5kg
Chemical Name or Material Potassium Sulfate