Potassium tantalum oxide, 99.998% (metals basis), Alfa Aesar™ Puratronic™

Manufacturer: Alfa Aesar 1287906

Quantity 5g

 View more versions of this product

Catalog No. AA1287906

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 12030-91-0
    Physical Form Powder
    Assay Percent Range 99.998% (metals basis)
    Molecular Formula KTaO3
    Formula Weight 268.05
    MDL Number MFCD00049652
    Synonym Potassium tantalate
    Chemical Name or Material Potassium tantalum oxide

    Specifications

    Formula Weight 268.05
    Quantity 5g
    Color White
    Physical Form Powder
    Odor Odorless
    Assay Percent Range 99.998% (metals basis)
    Molecular Formula KTaO3
    Appearance White powder
    MDL Number MFCD00049652
    CAS 12030-91-0
    Synonym Potassium tantalate
    Chemical Name or Material Potassium tantalum oxide

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit