Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Potassium tert-butoxide, 98+%, pure, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
2.5 kg
Packaging:
Glass Bottle
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 240.0°C to 258.0°C |
Color | White |
Assay Percent Range | 98+% |
Packaging | Glass Bottle |
Linear Formula | (CH3)3COK |
Quantity | 5 g |
Fieser | 01,911; 02,336; 03,233; 04,399; 05,544; 06,477; 08,407; 09,380; 10,323; 11,432; 12,97; 14,264; 17,289 |
Solubility Information | Solubility in water: reacts with water. Other solubilities: soluble in methanol, ethanol, slightly soluble in diethyl ether, benzene |
Formula Weight | 112.21 |
Percent Purity | 98+% |
Show More |
Safety and Handling
Hazard Category | GHS Signal Word: Danger |
Hazard Statement | GHS H Statement Causes severe skin burns and eye damage. Flammable solid. Self-heating in large quantities; may catch fire. Reacts violently with water. |
Precautionary Statement | GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Wear eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF IN EYES: Rinse cautiously with water for several min |
EINECSNumber | 212-740-3 |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction