Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium tert-butoxide, pure, 1M solution in THF, AcroSeal™, Acros Organics

Click to view available options
Quantity:
100mL
1L
2.5L
Packaging:
AcroSeal™ Glass Bottle
865-47-4,109-99-9
C4H9KO
112.21
LPNYRYFBWFDTMA-UHFFFAOYSA-N
potassium tert-butoxide,potassium tert-butanolate,potassium t-butoxide,potassium 2-methylpropan-2-olate,potassium tert-butylate,kotbu,2-methyl-2-propanol, potassium salt,tert-butoxypotassium,potassium-t-butoxide,t-buok
23665647
potassium;2-methylpropan-2-olate
CC(C)(C)[O-].[K+]
865-47-4,109-99-9
C4H9KO
112.21
LPNYRYFBWFDTMA-UHFFFAOYSA-N
potassium tert-butoxide,potassium tert-butanolate,potassium t-butoxide,potassium 2-methylpropan-2-olate,potassium tert-butylate,kotbu,2-methyl-2-propanol, potassium salt,tert-butoxypotassium,potassium-t-butoxide,t-buok
23665647
potassium;2-methylpropan-2-olate
CC(C)(C)[O-].[K+]
Linear Formula (CH3)3COK
Fieser 01,911; 02,336; 03,233; 04,399; 05,544; 06,477; 08,407; 09,380; 10,323; 11,432; 12,97; 14,264; 17,289
Formula Weight 112.21
Physical Form Solution
Grade Pure
Concentration 0.95 to 1.10M
Density 0.9020g/mL
Packaging AcroSeal™ Glass Bottle
Specific Gravity 0.902
Color Colorless to Orange-Yellow
Quantity 1L
Flash Point −19°C
Chemical Name or Material Potassium tert-butoxide
Show More Show Less

GHS H Statement
May cause respiratory irritation.
Causes severe skin burns and eye damage.
May form explosive peroxides.
Highly flammable liquid and vapor.
Suspected of causing cancer.
Reacts violently with water.<br/

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye p

GHS Signal Word: Danger

EINECSNumber : 212-740-3