Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Potassium tert-pentyloxide, 14-18% w/v in cyclohexane

Coreactant for conversion of ketones to oxides

Manufacturer:  Alfa Aesar™ 4139406

Quantity 5g

 View more versions of this product

Catalog No. AA4139406


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

41233-93-6
126.24
UN2924
Potassium tert-pentyloxide
CH3CH2C(CH3)2OK
MFCD00064808
Potassium tert-amylate; Potassium 2-methyl-2-butoxide

Specifications

41233-93-6
CH3CH2C(CH3)2OK
MFCD00064808
Potassium tert-amylate; Potassium 2-methyl-2-butoxide
14 to 18% w/v in cyclohexane
−18° (0°F)
Potassium tert-pentyloxide
Liquid
126.24
UN2924
0.80g/mL
5g
Air & Moisture Sensitive

Safety and Handling

SDS & Safety Documents

GHS H Statement
H225-H314-H318-H336
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause drowsiness or dizziness.

GHS P Statement
P210-P280-P273-P305+P351+P338-P310-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid release to the environment.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit