Learn More
Potassium tert-pentyloxide, 25% w/w in toluene, Thermo Scientific Chemicals
Description
Potassium tert-pentyloxide is a strong, hindered base used in combination with lithium diisopropyl amide to generate a superbase for the selective metallation of the aralkyl system at the 6-position of protected estradiols. It is used in organic synthesis, agrochemicals, pharmaceuticals, colorants and aroma chemicals. It acts as a catalyst in polymerization and isomerizations.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Potassium tert-pentyloxide is a strong, hindered base used in combination with lithium diisopropyl amide to generate a superbase for the selective metallation of the aralkyl system at the 6-position of protected estradiols. It is used in organic synthesis, agrochemicals, pharmaceuticals, colorants and aroma chemicals. It acts as a catalyst in polymerization and isomerizations.
Notes
Air and moisture sensitive. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents and acids. Reacts violently with water.
Specifications
Density | 0.87 |
Flash Point | 4°C (39°F) |
Quantity | 250 mL |
UN Number | UN2924 |
Beilstein | 4149025 |
Sensitivity | Air and moisture sensitive |
Formula Weight | 126.24 |
Concentration or Composition (by Analyte or Components) | 25% w/w in toluene |
Chemical Name or Material | Potassium tert-pentyloxide |
Safety and Handling
Hazard Category | H225-H304-H314-H335-H336-H361d-H373 |
Hazard Statement | GHS H Statement H225-H304-H361-H373-H314-H336 Highly flammable liquid and vapor. May be fatal if swallowed and enters airways. Suspected of damaging fertility or the unborn child. May cause damage to organs through prolonged or repeated exposure. Causes severe skin burns and eye damage. May cause drowsiness or dizziness. |
Precautionary Statement | P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c |
DOTInformation | Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S. |
EINECSNumber | 255-272-5 |
TSCA | Yes |
Recommended Storage | Ambient temperatures; Store under Argon |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.