Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium tetrabromoaurate(III) dihydrate, 99.99% (metals basis), Au 33% min., Alfa Aesar™ Premion™

Manufacturer:  Alfa Aesar 1266406

Quantity 5g

 View more versions of this product

Catalog No. AA1266406


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

14323-32-1
99.99% (metals basis)
591.72 (555.69 Anhydrous)
Gold potassium bromide
Crystalline
KAuBr4·2H2O
MFCD00049650
Potassium tetrabromoaurate(III) dihydrate, Au 33% min.

Specifications

14323-32-1
99.99% (metals basis)
Red-brown crystals
MFCD00049650
Weak
Red-brown
Potassium tetrabromoaurate(III) dihydrate, Au 33% min.
Crystalline
KAuBr4·2H2O
591.72 (555.69 Anhydrous)
Gold potassium bromide
Dihydrate
5g

Safety and Handling

SDS & Safety Documents

In case of contact with eyes, rinse immediately with plenty of water and seek medical advice.

H315-H319-H335: Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

EINECSNumber : 238-268-8

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit