Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium tetrachloroaurate(III) hydrate, 97%, Acros Organics

Click to view available options
Quantity:
1g
27988-75-6
AuCl4H2KO
395.88
MFCD00272631
BGVPYGKVUIFYCE-UHFFFAOYSA-J
potassium tetrachloroaurate iii hydrate,gold potassium chloride,aucl4.k.h2o,potassium hydrate tetrachloroaurate,potassium tetrachloroaurate iii hydrate, premion
91886546
potassium;tetrachlorogold(1-);hydrate
O.Cl[Au-](Cl)(Cl)Cl.[K+]
27988-75-6
AuCl4H2KO
395.88
MFCD00272631
BGVPYGKVUIFYCE-UHFFFAOYSA-J
potassium tetrachloroaurate iii hydrate,gold potassium chloride,aucl4.k.h2o,potassium hydrate tetrachloroaurate,potassium tetrachloroaurate iii hydrate, premion
91886546
potassium;tetrachlorogold(1-);hydrate
O.Cl[Au-](Cl)(Cl)Cl.[K+]
Assay Percent Range 97%
Linear Formula KAuCl4·xH2O
Formula Weight 377.87
Physical Form Crystals or Crystalline Powder
Percent Purity ≥96.0% (ex Au)
Color Orange
Quantity 1g
Chemical Name or Material Potassium tetrachloroaurate(III) hydrate