Potassium tetrachloropalladate(II), 99.99% (metals basis), Pd 32.2% min., Alfa Aesar™ Premion™

Manufacturer: Alfa Aesar 1188509

Quantity 10g

 View more versions of this product

Catalog No. AA1188509

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 10025-98-6
    Physical Form Crystalline
    Assay Percent Range 99.99% (metals basis)
    Molecular Formula K2PdCl4
    Formula Weight 326.42
    MDL Number MFCD00011373
    Chemical Name or Material Potassium tetrachloropalladate(II), Pd 32.2% min.

    Specifications

    Formula Weight 326.42
    Quantity 10g
    Color Red-brown
    EINECS Number 233-049-3
    Physical Form Crystalline
    Odor Odorless
    Assay Percent Range 99.99% (metals basis)
    Molecular Formula K2PdCl4
    Appearance Red-brown crystals
    MDL Number MFCD00011373
    CAS 10025-98-6
    Health Hazard 1 H290-H319-H302: May be corrosive to metals. Causes serious eye irritation. Harmful if swallowed.
    Chemical Name or Material Potassium tetrachloropalladate(II), Pd 32.2% min.

    H290-H319-H302: May be corrosive to metals. Causes serious eye irritation. Harmful if swallowed.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit