Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium tetrachloropalladate(II), min 32.0% Pd, ACROS Organics™

Click to view available options
Quantity:
2g
10g
Packaging:
Glass bottle
10025-98-6
Cl4K2Pd
326.41
MFCD00011373
LGCKLDWLSVFMGL-UHFFFAOYSA-J
potassium tetrachloropalladate ii,potassium chloropalladite,potassium tetrachloropalladate,ccris 6625,potassium chlorophalladite,potassium palladium chloride,potassium palladous chloride,dipotassium tetrachloropalladate ii,dipotassium tetrachloropalladate 2-,tetrachloropalladate 2-dipotassium
61438
dipotassium;tetrachloropalladium(2-)
Cl[Pd-2](Cl)(Cl)Cl.[K+].[K+]
10025-98-6
Cl4K2Pd
326.41
MFCD00011373
LGCKLDWLSVFMGL-UHFFFAOYSA-J
potassium tetrachloropalladate ii,potassium chloropalladite,potassium tetrachloropalladate,ccris 6625,potassium chlorophalladite,potassium palladium chloride,potassium palladous chloride,dipotassium tetrachloropalladate ii,dipotassium tetrachloropalladate 2-,tetrachloropalladate 2-dipotassium
61438
dipotassium;tetrachloropalladium(2-)
Cl[Pd-2](Cl)(Cl)Cl.[K+].[K+]
Linear Formula K2PdCl4
Solubility Information Solubility in water: soluble. Other solubilities: insoluble in alcohol
Formula Weight 326.41
Physical Form Crystals or Crystalline Powder
Percent Purity min 32% Pd
Calcium (Ca) 10ppm max.
Copper (Cu) 10ppm max.
Density 2.6700g/mL
Gold (Au) 20ppm max.
Iridium (Ir) 20ppm max.
Iron (Fe) 10ppm max.
Magnesium (Mg) 10ppm max.
Packaging Glass bottle
Platinum (Pt) 20ppm max.
Rhodium (Rh) 20ppm max.
Ruthenium (Ru) 20ppm max.
Silicon (Si) 30ppm max.
Silver (Ag) 20ppm max.
Solubility Solubility in water: soluble. Other solubilities: insoluble in alcohol
Specific Gravity 2.67
Color Brown to Brown-Green
Melting Point 105.0°C
Quantity 10g
Chemical Name or Material Potassium tetrachloropalladate(II), min 32.0% Pd
Show More Show Less

GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a

GHS Signal Word: Warning

EINECSNumber : 233-049-3

RTECSNumber : TT4100000

TSCA : TSCA