Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium tetrachloroplatinate, 99.99%, (trace metal basis), ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
10025-99-7
Cl4K2Pt
415.1
RVRDLMCWUILSAH-UHFFFAOYSA-J
potassium tetrachloroplatinate ii,dipotassium tetrachloroplatinate,potassium tetrachloroplatinate,potassium chloroplatinite,potassium tetrachloroplatinite,dipotassium tetrachloroplatinate 2-,potassium platinochloride,platinous potassium chloride,potassium platinous chloride,bipotassium tetrachloroplatinate
61440
dipotassium;tetrachloroplatinum(2-)
Cl[Pt-2](Cl)(Cl)Cl.[K+].[K+]
10025-99-7
Cl4K2Pt
415.1
RVRDLMCWUILSAH-UHFFFAOYSA-J
potassium tetrachloroplatinate ii,dipotassium tetrachloroplatinate,potassium tetrachloroplatinate,potassium chloroplatinite,potassium tetrachloroplatinite,dipotassium tetrachloroplatinate 2-,potassium platinochloride,platinous potassium chloride,potassium platinous chloride,bipotassium tetrachloroplatinate
61440
dipotassium;tetrachloroplatinum(2-)
Cl[Pt-2](Cl)(Cl)Cl.[K+].[K+]
for ultra trace analysis
Assay Percent Range 99.99%
Linear Formula K2PtCl4
Merck Index 15,7801
Solubility Information Solubility in water: 10g/L (20°C)
Formula Weight 415.1
Physical Form Crystals
Percent Purity 99.99%
Grade Trace Metal Basis
Packaging Glass bottle
Solubility Solubility in water: 10g/L (20°C).
Total Trace Metal Impurities 100ppm max.
Color Brown or Orange to Red
Melting Point 250.0°C
Quantity 1g
Chemical Name or Material Potassium tetrachloroplatinate
Show More Show Less

GHS H Statement:
Toxic if swallowed.
Causes skin irritation.
Causes serious eye damage.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.

GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 233-050-9