Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium tetraiodomercurate(II) 99.99%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
7783-33-7
HgI4K2
786.406
MFCD00049656
OPCMAZHMYZRPID-UHFFFAOYSA-J
nessler's reagent,nessler reagent,mayer's reagent,channing's solution,caswell no. 695,potassium iodomercurate,mercury ii potassium iodide,mercury potassium iodide,potassium mercuric iodide,mercuric potassium iodide
24542
CHEBI:51568
dipotassium;tetraiodomercury(2-)
[K+].[K+].I[Hg-2](I)(I)I
7783-33-7
HgI4K2
786.406
MFCD00049656
OPCMAZHMYZRPID-UHFFFAOYSA-J
nessler's reagent,nessler reagent,mayer's reagent,channing's solution,caswell no. 695,potassium iodomercurate,mercury ii potassium iodide,mercury potassium iodide,potassium mercuric iodide,mercuric potassium iodide
24542
CHEBI:51568
dipotassium;tetraiodomercury(2-)
[K+].[K+].I[Hg-2](I)(I)I
For ultra trace analysis
Assay Percent Range 99.99%
Linear Formula K2HgI4
Merck Index 15, 7805
Formula Weight 786.39
Physical Form Crystals To Granules
Grade Trace Metal Basis
Packaging Glass bottle
Total Trace Metal Impurities 100ppm max.
Color Yellow to Orange
Quantity 5g
Chemical Name or Material Potassium tetraiodomercurate(II)
Show More Show Less

GHS H Statement:
Fatal in contact with skin.
Fatal if swallowed.
May cause damage to organs through prolonged or repeated exposure.
Fatal if inhaled.
Very toxic to aquatic life with long lasting effects.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Remove/Take off immediately all contaminated clothing.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger