Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium thiocyanate, 99+%, for analysis, ACROS Organics™

Click to view available options
Quantity:
250g
500g
2.5kg
10kg
Packaging:
Plastic bottle
Plastic drum
333-20-0
CKNS
97.18
MFCD00011413
ZNNZYHKDIALBAK-UHFFFAOYSA-M
potassium thiocyanate,potassium rhodanate,potassium rhodanide,rodanca,potassium sulfocyanate,rhodanide,rhocya,potassium thiocyanide,kscn,thiocyanic acid, potassium salt
516872
CHEBI:30951
potassium;thiocyanate
C(#N)[S-].[K+]
333-20-0
CKNS
97.18
MFCD00011413
ZNNZYHKDIALBAK-UHFFFAOYSA-M
potassium thiocyanate,potassium rhodanate,potassium rhodanide,rodanca,potassium sulfocyanate,rhodanide,rhocya,potassium thiocyanide,kscn,thiocyanic acid, potassium salt
516872
CHEBI:30951
potassium;thiocyanate
C(#N)[S-].[K+]
Indicator
Assay Percent Range 99% min. (Argentometry)
Linear Formula KSCN
Ammonium 10ppm max.
Fieser 01,954
Merck Index 15, 7807
Solubility Information Solubility in water: 2170g/L (20°C). Other solubilities: soluble in alcohol and acetone
Formula Weight 97.18
Physical Form Crystalline Powder
Percent Purity 99+%
Grade Analysis
Chloride 50ppm max
Chlorine (Cl) 50ppm max.
Density 1.8860g/mL
Heavy Metals (as Pb) 2ppm max.
Iodine Consuming Subst. 30mg I2/100g max.
Iron (Fe) 2ppm max.
Packaging Plastic drum
Sodium (Na) 200ppm max.
Solubility Solubility in water: 2170g/L (20°C). Other solubilities: soluble in alcohol and acetone
Specific Gravity 1.886
Sulfate 50ppm max.
Color White
Melting Point 170.0°C to 179.0°C
pH 5.3 to 8.7 (5% soln. at 20°C)
Quantity 10kg
Chemical Name or Material Potassium thiocyanate, p.a.
Show More Show Less

GHS H Statement:
Harmful to aquatic life with long lasting effects.
Harmful if swallowed.
Harmful if inhaled.
Harmful in contact with skin.
Contact with acids liberates very toxic gas.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid release to the environment.
Keep container tightly closed.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 206-370-1

RTECSNumber : XL1925000

TSCA : TSCA