Potassium thiocyanate, ACS, 98.5% min, Alfa Aesar™

Manufacturer: Alfa Aesar 1431822

Quantity 100g

 View more versions of this product

Catalog No. AA1431822

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 333-20-0
    Physical Form Crystals
    Assay Percent Range 98.5% min.
    Molecular Formula KSCN
    Formula Weight 97.18
    MDL Number MFCD00011413
    Synonym Potassium sulfocyanate
    Chemical Name or Material Potassium thiocyanate, For ACS analysis
    Grade ACS Grade

    Specifications

    Physical Form Crystals
    Formula Weight 97.18
    Quantity 100g
    Color White
    Density 1.89g/cm3
    EINECS Number 206-370-1
    Odor Odorless
    Assay Percent Range 98.5% min.
    Molecular Formula KSCN
    Appearance White crystalline
    MDL Number MFCD00011413
    CAS 333-20-0
    Synonym Potassium sulfocyanate
    Health Hazard 1 H302-H312-H332-H412: Harmful if swallowed. Harmful in contact with skin. Harmful if inhaled. Harmful to aquatic life with long lasting effects.
    Chemical Name or Material Potassium thiocyanate, For ACS analysis
    Grade ACS Grade

    H302-H312-H332-H412: Harmful if swallowed. Harmful in contact with skin. Harmful if inhaled. Harmful to aquatic life with long lasting effects.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit