Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Honeywell™ Potassium thiocyanate, Honeywell
Click to view available options
Quantity:
250g
1kg
2.5kg
Description
- Products with a guarantee certificate and/or suitable for the stated analytical application.
Specifications
Linear Formula | KSCN |
UN Number | NONH for all modes of transport |
Beilstein | 3594799 |
Formula Weight | 97.18g/mol |
Percent Purity | ≥98% (AT) |
Quantity | 1kg |
Chemical Name or Material | Potassium thiocyanate |
Safety and Handling
P273-P280
RTECSNumber : XL1925000