Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Potassium thiocyanate, Honeywell™
Click to view available options
Quantity:
1 kg
2.5 kg
Description
- Products with a guarantee certificate and/or suitable for the stated analytical application.
Specifications
Quantity | 2.5 kg |
Linear Formula | KSCN |
UN Number | NONH for all modes of transport |
Beilstein | 3594799 |
Formula Weight | 97.18g/mol |
Percent Purity | ≥98% (AT) |
Chemical Name or Material | Potassium thiocyanate |
Safety and Handling
P273-P280
RTECSNumber : XL1925000