Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium thioglycolate, 40-44 wt.% solution in water, ACROS Organics™

Click to view available options
Quantity:
250mL
1L
Packaging:
Glass bottle
34452-51-2,7732-18-5
C2H3KO2S
130.202
MFCD00067265
HYTYHTSMCRDHIM-UHFFFAOYSA-M
potassium thioglycolate,unii-xp5wzo5w6h,potassium 2-mercaptoacetate,acetic acid, mercapto-, monopotassium salt,xp5wzo5w6h,68-11-1 parent,potassium sulfanylacetate,potassium mercaptoacetate,kopcolate,potassium ion thioglycolate
23688957
potassium;2-sulfanylacetate
C(C(=O)[O-])S.[K+]
34452-51-2,7732-18-5
C2H3KO2S
130.202
MFCD00067265
HYTYHTSMCRDHIM-UHFFFAOYSA-M
potassium thioglycolate,unii-xp5wzo5w6h,potassium 2-mercaptoacetate,acetic acid, mercapto-, monopotassium salt,xp5wzo5w6h,68-11-1 parent,potassium sulfanylacetate,potassium mercaptoacetate,kopcolate,potassium ion thioglycolate
23688957
potassium;2-sulfanylacetate
C(C(=O)[O-])S.[K+]
Density 1.2500g/mL
Formula Weight 130.21
Physical Form Solution
Percent Purity 40.0 to 44.0%
Identification potassium: positive, mercaptane: positive
Iron (Fe) 0.5ppm max.
Packaging Glass bottle
Solubility Solubility in water: soluble.
Specific Gravity 1.25
Color Pink
pH 6.8 to 7.3
Quantity 250mL
Chemical Name or Material Potassium thioglycolate, 40-44 wt.% solution in water
Show More Show Less

GHS H Statement:
Harmful if swallowed.
May cause an allergic skin reaction.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning