Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium thiosulfate hydrate, 96+%, extra pure, Acros Organics

Click to view available options
Quantity:
100g
500g
Packaging:
Plastic bottle
1264198-56-2
K2O3S2·xH2O
190.33
MFCD00150405
FGRVOLIFQGXPCT-UHFFFAOYSA-L
61501
dipotassium;dioxido-oxo-sulfanylidene-$l^{6}-sulfane
[O-]S(=O)(=S)[O-].[K+].[K+]
1264198-56-2
K2O3S2·xH2O
190.33
MFCD00150405
FGRVOLIFQGXPCT-UHFFFAOYSA-L
61501
dipotassium;dioxido-oxo-sulfanylidene-$l^{6}-sulfane
[O-]S(=O)(=S)[O-].[K+].[K+]
Reduction reagent
Assay Percent Range 96+%
Linear Formula K2S2O3·xH2O
Solubility Information Solubility in water: soluble. Other solubilities: insoluble in alcohol
Formula Weight 190.33
Physical Form Liquid
Percent Purity ≥96%
Grade Extra Pure
Chloride 0.05% max.
Heavy Metals (as Pb) 10ppm max.
Insoluble in Water 0.05% max.
Iron (Fe) 0.001% max.
Loss on Drying 4% max. (150°C)
Packaging Plastic bottle
Solubility Solubility in water: soluble. Other solubilities: insoluble in alcohol
Color Colorless to Yellow
pH 6 to 9 (5% soln.)
Quantity 500g
Chemical Name or Material Potassium thiosulfate hydrate
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning