Alfa Aesar™ Potassium titanium oxide, 98% (metals basis)

Manufacturer: Alfa Aesar™ 1413822

Quantity 100g

 View more versions of this product

Catalog No. AA1413822

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 12030-97-6
    Formula Weight 174.06
    MDL Number MFCD00058803
    Chemical Name or Material Potassium titanium oxide

    Specifications

    Formula Weight 174.06
    TSCA TSCA
    Mesh Size −325 Mesh
    Odor Odorless
    Physical Form Powder
    MDL Number MFCD00058803
    CAS 12030-97-6
    Melting Point 1615°C
    Quantity 100g
    Chemical Name or Material Potassium titanium oxide
    Assay 98%, (metals basis)

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit