Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium triisopropoxyborohydride, 1M solution in THF, AcroSeal™, ACROS Organics™

Click to view available options
Quantity:
100mL
800mL
Packaging:
AcroSeal™ Glass Bottle
AcroSeal™ Glass Bottle in Tin Can
42278-67-1,109-99-9
C9H22BKO3
228.18
MFCD00044439
XMTTXBQMYHBFNI-UHFFFAOYSA-N
potassium hydro triisopropoxy borate,potassium triisopropoxyhydroborate,potassium triisopropoxyborohydride1,potassium triisopropoxy-? 2-boranuide,potassium triisopropoxyborohydride in tetrahydrofuran
42278-67-1,109-99-9
C9H22BKO3
228.18
MFCD00044439
XMTTXBQMYHBFNI-UHFFFAOYSA-N
potassium hydro triisopropoxy borate,potassium triisopropoxyhydroborate,potassium triisopropoxyborohydride1,potassium triisopropoxy-? 2-boranuide,potassium triisopropoxyborohydride in tetrahydrofuran
Reduction reagent
Linear Formula KB[OCH(CH3)2]3H
Fieser 09,393; 12,414
Density 0.9140g/mL
Formula Weight 228.18
Physical Form Solution
Percent Purity 1M (exact strength on the certificate of analysis)
Packaging AcroSeal™ Glass Bottle
Color Colorless to Yellow
Quantity 800mL
Chemical Name or Material Potassium triisopropoxyborohydride, 1M solution in THF

GHS P Statement Causes severe skin burns and eye damage. May cause respiratory irritation. In contact with water releases flammable gases which may ignite spontaneously. Highly flammable liquid and vapour. Reacts viol

GHS P Statement Keep away from any possible contact with water, because of violent reaction and possible flash fire. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiou

Danger