Alfa Aesar™ Potassium tungsten oxide, 99.5% (metals basis)

Manufacturer: Alfa Aesar™ 1403122

Quantity 100g

 View more versions of this product

Catalog No. AA1403122

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7790-60-5
    Formula Weight 326.04
    MDL Number MFCD00049668
    Merck Index 14,7694
    Synonym Potassium tungstate
    Chemical Name or Material Potassium tungsten oxide

    Specifications

    Formula Weight 326.04
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 2 GHS H Statement
    H319-H335
    Causes serious eye irritation.
    May cause respiratory irritation.
    Odor Odorless
    Physical Form Powder
    MDL Number MFCD00049668
    CAS 7790-60-5
    Melting Point 921°C
    Quantity 100g
    Solubility Information Soluble in water
    Sensitivity Hygroscopic
    Merck Index 14,7694
    Synonym Potassium tungstate
    Health Hazard 1 Warning
    Density 3.12g/mL
    TSCA TSCA
    Mesh Size −60 Mesh
    Chemical Name or Material Potassium tungsten oxide
    Assay 99.5%, (metals basis)

    GHS H Statement
    H319-H335
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit