Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Potassium vinyltrifluoroborate, 95%, Acros Organics
Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
Specifications
Melting Point | 240°C to 242°C |
Color | White to Yellow |
Packaging | Glass bottle |
Quantity | 5g |
Infrared Spectrum | Authentic |
Assay Percent Range | 94% min |
Formula Weight | 133.95 |
Physical Form | Powder |
Percent Purity | 95% |
Grade | Technical |
Show More |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
GHS Signal Word: Danger