Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ POTEH Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA524910
Description
POTEH Polyclonal Antibody for Western Blot, Flow
POTEH belongs to the POTE family and contains 7 ANK repeats.
Specifications
POTEH | |
Polyclonal | |
PBS with 0.09% sodium azide; pH 7.4 | |
Q6S545 | |
POTEH | |
KLH conjugated synthetic peptide between 32-60 amino acids from the N-terminal region of human POTEH. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Flow Cytometry, Western Blot | |
Unconjugated | |
POTEH | |
A26C3; ACTBL1; actin, beta-like 1; ANKRD26-like family C member 3; ANKRD26-like family C, member 3; cancer/testis antigen family 104, member 7; CT104.7; POTE ankyrin domain family member H; POTE ankyrin domain family, member H; POTE22; POTE-22; POTEH; Prostate, ovary, testis-expressed protein on chromosome 22; protein expressed in prostate, ovary, testis, and placenta 22; protein expressed in prostate, ovary, testis, and placenta POTE14 like | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
23784 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction