Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

POTEH Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$106.19 - $352.89

Specifications

Antigen POTEH
Dilution Western Blot 1.0 ug/ml
Applications Western Blot
Conjugate Unconjugated
Format Purified
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB124144
View Documents
Novus Biologicals
NBP30962125UL
25 μg
Each for $106.19
Add to cart
 
NB124143
View Documents
Novus Biologicals
NBP309621100UL
100 μg
Each for $352.89
Add to cart
 
Description

Description

POTEH Polyclonal antibody specifically detects POTEH in Human samples. It is validated for Western Blot
Specifications

Specifications

POTEH
Western Blot
Purified
RUO
PBS buffer, 2% sucrose
23784
Polyclonal
Affinity purified
Western Blot 1.0 ug/ml
Unconjugated
Rabbit
Human
A26C3, ACTBL1ANKRD26-like family C member 3, actin, beta-like 1, ANKRD26-like family C, member 3, cancer/testis antigen family 104, member 7, CT104.7, POTE ankyrin domain family member H, POTE ankyrin domain family, member H, POTE22POTE-22, Prostate, ovary, testis-expressed protein on chromosome 22, protein expressed in prostate, ovary, testis, and placenta 22, protein expressed in prostate, ovary, testis, and placenta POTE14 like
The immunogen is a synthetic peptide directed towards the C-terminal region of Human POTEH (NP_001129685). Peptide sequence EEESQRLKGSENSQPEEMSQEPEINKGGDRKVEEEMKKHGSTHMGFPENL
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit