Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

POTEM Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549491

Catalog No. PA549491


Add to cart

Specifications

Specifications

POTEM
Polyclonal
PBS with 0.09% sodium azide
A6NI47
Putative POTE ankyrin domain family member M, POTEM
Rabbit
IgG
50 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
641455
Western Blot
Unconjugated
POTEM
Liquid
POTEM
KLH conjugated synthetic peptide between 430-462 amino acids from the C-terminal region of human POTEM.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit