Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

POU2F3 Antibody, Novus Biologicals™
SDP

Goat Polyclonal Antibody

Supplier:  Novus Biologicals NBP149870

Catalog No. NBP149870


Only null left
Add to Cart

Description

Description

POU2F3 Polyclonal specifically detects POU2F3 in Human samples. It is validated for Western Blot, Peptide ELISA.
Specifications

Specifications

POU2F3
Polyclonal
Western Blot 1 - 3 ug/ml, Peptide ELISA Detection limit 1:32000
NP_055167.2
POU2F3
Peptide with sequence C-EPHLEASQHLPVPKH corresponding to internal region according to NP_055167.2.
0.1 mg
Primary
This antibody recognizes and epitope within the internal region.This antibody is specific to the internal region. Cross reacts with Human. Expected from sequence similarity: Rat, Dog and Cow.
Store at -20C. Avoid freeze-thaw cycles.
Western Blot, ELISA
Unconjugated
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Epoc-1, MGC126698, oct-11, OCT11FLJ40063, Octamer-binding protein 11, Octamer-binding transcription factor 11, OTF11, OTF-11, PLA1, PLA-1, POU class 2 homeobox 3, POU domain class 2, transcription factor 3, POU domain, class 2, transcription factor 3, POU transcription factor, Skn-1a, Transcription factor PLA-1, Transcription factor Skn-1
Goat
Affinity Purified
RUO
25833
Human
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

For Research Use Only