Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
POU3F3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 189991AP
Catalog No. 50-173-2315
Description
Description
The antibody is specific to OTF8.
POU3F3 is a member of the class III POU family of transcription factors that are expressed in the central nervous system. The POU domain in these proteins is required for high affinity binding to octamer DNA sequences.
Specifications
Specifications
POU3F3
0.23 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P20264, P31361, Q63262
Brain 1, Brn 1, BRN1, Oct 8, Octamer binding protein 8, OTF 8, OTF8, POU class 3 homeobox 3, POU3F3
Rabbit
IgG
150 μL
-20°C
Primary
18993, 192109, 5455
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
POU3F3
Liquid
Pou3f3
Peptide
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat