Learn More
POU3F4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier:  Thermo Scientific PA124479
Catalog No. PIPA124479
Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative.
Description
Description
PA1-24479 detects POU3F4 in Human samples. PA1-24479 has been successfully used in Western Blot procedures. PA1-24479 immunogen corresponds to Polyclonal antibody from rabbits immunized with a synthetic peptide region of human POU3F4. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.
POU3F4 (brain-specific homeobox/POU domain protein 4, brain-4, Brn-4, transcription factor 4) is a transcription factor with a POU domain.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
POU3F4
Polyclonal
Unconjugated
P49335, P62515, P62516
POU3F4
Synthetic peptide region of human POU3F4.
100 μg
Primary
Canine, Human, Mouse, Rat
Antibody
IgG
Western Blot
1 mg/mL
POU3F4
BRAIN-4, BRN4, DFN3, DFNX2, OTF9
Rabbit
IgG fraction
RUO
18994, 29589, 491988, 5456
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Recommendations
Recommendations